Learn More
LASS2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
136.48CHF - 385.92CHF
Spécification
Antigène LASS2
Concentration 0.21 mg/mL
Applications Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugué Unconjugated
Consulter d'autres spécifications
Produits 2
Code produit Marque Quantité Prix Quantité et disponibilité  
Code produit Marque Quantité Prix Quantité et disponibilité  
16819874
Afficher les documents
Proteintech
20344-1-AP-20UL
20 μL
136.48CHF
20µL
Il en reste null
En stock
Ajouter au panier
16809874
Afficher les documents
Proteintech
20344-1-AP-150UL
150 μL
385.92CHF
150µL
Il en reste null
En stock
Ajouter au panier
Description
Description
LASS2 is a protein that has sequence similarity to yeast longevity assurance gene 1. Mutation or overexpression of the related gene in yeast has been shown to alter yeast lifespan. The human protein may play a role in the regulation of cell growth. This gene encodes a protein that has sequence similarity to yeast longevity assurance gene 1. Mutation or overexpression of the related gene in yeast has been shown to alter yeast lifespan. The human protein may play a role in the regulation of cell growth. Alternatively spliced transcript variants encoding the same protein have been described.
Spécification
Spécification
LASS2
Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
Human, Mouse
Q924Z4, Q96G23
29956, 76893
LASS2 Fusion Protein Ag14151
Primary
-20°C
CERS2
0.21 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
CerS2, L3, LASS2, SP260, TMSG1
Cers2
IgG
Antigen Affinity Chromatography