Learn More
LASS2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 20344-1-AP-150UL
 Afficher plus de versions de ce produit
Code produit. 16809874
  • 410.00€ / 150µL
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
LASS2 is a protein that has sequence similarity to yeast longevity assurance gene 1. Mutation or overexpression of the related gene in yeast has been shown to alter yeast lifespan. The human protein may play a role in the regulation of cell growth. This gene encodes a protein that has sequence similarity to yeast longevity assurance gene 1. Mutation or overexpression of the related gene in yeast has been shown to alter yeast lifespan. The human protein may play a role in the regulation of cell growth. Alternatively spliced transcript variants encoding the same protein have been described.
Spécification
Spécification
LASS2
Polyclonal
Unconjugated
CERS2
CerS2, L3, LASS2, SP260, TMSG1
Rabbit
Antigen Affinity Chromatography
RUO
29956, 76893
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q924Z4, Q96G23
Cers2
LASS2 Fusion Protein Ag14151
150 μL