Chromatografie-instrumenten
Thermo Scientific™ Acclaim™ Guard Holder and Coupler
Use the Thermo Scientific™ Guard Holder and Coupler for your Thermo Scientific™ Acclaim™ Guard columns. They can be purchased separately or as a kit.
SCAT Safety Caps
The SCAT Safety Caps provide maximum safety for your HPLC solvent supply. Extract your solvents safely from bottles and reservoirs without being exposed to hazardous gases and solvent vapours. Complete with integrated air valve for pressure equalisation.
Thermo Scientific™ Dionex™ cytochroom C-digestaat
Thermo Scientific™ Dionex Cytochrome C Digest is lyophilized.
Thermo Scientific™ Injection Port Liners for Agilent GC Instruments
Use these injection port liners to optimize performance of Agilent GC instruments.
Thermo Scientific™ pH Gradient Buffers
Robust reproducible gradients that are applicable to a wide range of MAbs and ready to use with existing LC columns and systems, without the need to formulate mobile phases.
Thermo Scientific™ Valco Injector, Model C6W
Valco continues to offer a diverse line in terms of number of ports, fitting sizes, and materials of construction.
Thermo Scientific™ RheBuild™ Kit
Eliminate the need to order individual parts. Included in each individualized RheBuild Kit are all the parts, tools, and instructions to maintain the precision performance of Rheodyne stainless steel and PEEK platforms.
Thermo Scientific™ Bottom-of-the-Bottle™ Solvent Filters
Protect your HPLC system with Thermo Scientific™ Bottom-of-the-Bottle™ Solvent Filters.
Thermo Scientific™ LinerGOLD™ GC Liners
Use the gold-standard LinerGOLD; GC Liners for consistently high accuracy, sensitivity, and precision in your gas chromatography (GC) analysis.
Thermo Scientific™ POROS™ MabCapture™ A Select Resin
MabCapture A Select a monoclonal antibody purification resin provides high dynamic binding at low linear flow rates and maintains superior binding capacity at high linear flow rates
Kinesis™ Inlet Solvent Filters
Stainless-steel or multiple materials
Thermo Scientific™ Injection Port Liners with CarboFrit for Thermo Scientific™ Trace and Focus SSL Instruments
Ensure a high degree of reproducibility with Thermo Scientific Injection Port Liners with CarboFrit for Thermo Scientific Trace and Focus SSL Instruments.
Thermo Scientific™ Rheodyne™ Sample Loops
Thermo Scientific™ Rheodyne Sample Loops are for Rheodyne sample injectors in stainless steel or biocompatible PEEK.
Thermo Scientific™ POROS™ XQ Strong Anion Exchange Resin
POROS XQ is a high-capacity, high-resolution, salt-tolerant resin that enables >140 mg/mL dynamic binding capacity while delivering exceptional separation performance
Thermo Scientific™ Rheodyne™ Replacement Rotor Seals for Injectors
Maintain performance of your Thermo Scientific™ Rheodyne™ Sample Injectors for HPLC with these replacement rotor seals.
Thermo Scientific™ Rheodyne™ 7725 and 7725i Sample Injectors
Ensure accurate, precise HPLC injections and protect against pressure shock with the dual-mode Thermo Scientific™ Rheodyne™ 7725 and 7725i Sample Injectors.
Thermo Scientific™ POROS™ XS Strong Cation Exchange Resin
POROS XS is the first high-capacity, high-resolution resin that allows loading to >100 mg/mL in the presence of 15 mS/cm NaCl, while delivering unprecedented separation capability
Thermo Scientific™ Dionex™ PEEK Sleeves
Thermo Scientific™ Dionex PEEK Sleeves are precision cut and polished.
Thermo Scientific™ GC Injection Port Base Seals for Thermo Scientific™ and Agilent Instruments
Ensure optimal GC analysis by using the exceptional sealing and inert properties of Thermo Scientific™ GC Injection Port Base Seals.
Thermo Scientific™ POROS™ HQ 50 µm Strong Anion Exchange Resin
POROS XQ is a high-capacity, high-resolution, salt-tolerant resin that enables >140 mg/mL dynamic binding capacity while delivering exceptional separation performance
DWK Life Sciences Kimble™ Kontes™ Straight Low Hold Up without Tip O-Ring PTFE Plug Hi-VAC Valves
With bended side arms to minimize liquid holdup
Thermo Scientific™ TracePLOT Particle Traps for GC Instruments
Safeguard your GC instruments from damage caused by dislodged particles entering the detector.
Thermo Scientific™ GC Liner Sealing Rings for Thermo Scientific™ and Agilent Instruments
Match the best sealing ring with the injection port liner used in your Thermo Scientific™ or Agilent GC system.
Idex PEEK™ Sample Loops
Offer consistent volumes due to tight tolerances of extrusion processes
Thermo Scientific™ Valco™ Accessories
Maintain the performance of your Thermo Scientific Valco™ Injector for high-performance liquid chromatography (HPLC) with these Valco HPLC Injector Accessories, which include syringe ports, ferrules, and nuts.
Even geduld terwijl wij uw resultaten te halen