missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WWP2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 12197-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This gene encodes a member of the NEDD4-like protein family. The family of proteins is known to possess ubiquitin-protein ligase activity. The encoded protein contains 4 tandem WW domains. The WW domain is a protein motif consisting of 35 to 40 amino acids and is characterized by 4 conserved aromatic residues. The WW domain may mediate specific protein-protein interactions. Three alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.Spécification
WWP2 | |
Polyclonal | |
Unconjugated | |
WWP2 | |
AIP2, WW domain containing protein 2, WWP2, WWp2 like | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11060, 66894 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O00308, Q9DBH0 | |
Wwp2 | |
WWP2 Fusion Protein Ag2835 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu