Learn More
USP25 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 67769-1-IG-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The USP25 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human USP25. This antibody recognizes human, mouse, rat antigen. The USP25 antibody has been validated for the following applications: WB, IHC, ELISA analysis.Spécification
USP25 | |
Monoclonal | |
1000 μg/mL | |
USP21 | |
Recombinant Protein | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
125 kDa | |
Liquid |
Western Blot, Immunohistochemistry, ELISA | |
2B6A2 | |
Unconjugated | |
Mouse | |
Protein A purification | |
RUO | |
29761 | |
Store at -20°C. Stable for one year after shipment. | |
Monoclonal Antibody | |
IgG2b |