Learn More
SLC2A9 Mouse anti-Human, Unconjugated, Clone: 2D6E1, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 67530-1-IG-20UL
Description
The SLC2A9 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SLC2A9. This antibody recognizes human, mouse, rat, rabbit, pig antigen. The SLC2A9 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Spécification
SLC2A9 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig, Rabbit |
Western Blot, Immunohistochemistry, Immunofluorescence | |
2D6E1 | |
Unconjugated | |
20 μL | |
56606 | |
IgG1 |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.