Learn More
RENALASE Mouse anti-Human, Clone: 3A12F5, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 60128-1-IG-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This antibody is specific to RNLS.
Renalase is a flavin adenine dinucleotide-dependent amine oxidase that is secreted into the blood from the kidney.Spécification
RENALASE | |
Monoclonal | |
0.6 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5VYX0 | |
RNLS | |
RENALASE Fusion Protein Ag13061 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
3A12F5 | |
Unconjugated | |
RNLS | |
C10orf59, MAO C, Monoamine oxidase C, RENALASE, RNLS | |
Mouse | |
Protein G | |
RUO | |
55328 | |
-20°C | |
Liquid |