Learn More
PTPN23 Rabbit, anti-Human, CoraLite 647, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech CL647-10472-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The PTPN23 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PTPN23. This antibody recognizes human antigen. The PTPN23 antibody has been validated for the following applications: IF analysis. This antibody is coralite™647.
Spécification
PTPN23 | |
1000 μg/mL | |
DKFZp564F0923, HD PTP, HDPTP, KIAA1471, PTP TD14, PTPN23 | |
20 μL | |
25930 | |
Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
Liquid |
Immunofluorescence | |
CoraLite 647 | |
Rabbit | |
Primary | |
Human | |
150 kDa | |
IgG |