Learn More
Sampling Systems™ Flacons PTFE
PTFE bottles are ideal for storage and are made from 100% PTFE, giving them high chemical resistance. They are suitable for low temperatures and include both a bottle and a cap.
Marque: Sampling Systems™ A125-10
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
- Conforms to both FDA and USP class VI requirements
- Crevice-free interior
- Heavy wall construction
- Suitable for use up to 280°C
Spécification
10 mL | |
White | |
Non stérile | |
Autoclavable | |
No | |
Cylindrique | |
Storage Bottle | |
PTFE | |
Oui |
0.34 oz. | |
PTFE | |
Oui | |
Reusable | |
Large | |
Certified Quality | |
Unlined | |
Screw Cap |