missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEIS2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 11550-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This gene encodes a homeobox protein belonging to the TALE (′three amino acid loop extension′) family of homeodomain-containing proteins. TALE homeobox proteins are highly conserved transcription regulators, and several members have been shown to be essential contributors to developmental programs. Multiple transcript variants encoding distinct isoforms have been described for this gene.Spécification
MEIS2 | |
Polyclonal | |
Unconjugated | |
MEIS2 | |
Homeobox protein Meis2, HsT18361, Meis homeobox 2, Meis1 related protein 1, MEIS2, MRG1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17536, 311311, 4212 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O14770, P97367 | |
Meis2 | |
MEIS2 Fusion Protein Ag2096 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu