Learn More
Importin Beta Mouse anti-Human, Unconjugated, Clone: 1A12B1, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 67597-1-IG-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The Importin Beta antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Importin Beta. This antibody recognizes human antigen. The Importin Beta antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spécification
Importin Beta | |
Monoclonal | |
780 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, ELISA | |
1A12B1 | |
Unconjugated | |
150 μL | |
3837 | |
IgG2b |