missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFFO1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
136.48CHF - 385.92CHF
Specifications
Antigen | IFFO1 |
---|---|
Concentration | 0.52 mg/mL |
Applications | Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
This antibody works well in WB, IHC and IF applications.
This gene is a member of the intermediate filament family. Intermediate filaments are proteins which are primordial components of the cytoskeleton and nuclear envelope. The proteins encoded by the members of this gene family are evolutionarily and structurally related but have limited sequence homology, with the exception of the central rod domain. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
IFFO1 | |
Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q0D2I5, Q8BXL9 | |
25900, 320678, 362437 | |
IFFO1 Fusion Protein Ag8915 | |
Primary | |
-20°C | |
IFFO1 |
0.52 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
HOM TES 103, IFFO, IFFO1, Tumor antigen HOM TES 103 | |
Iffo1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title