missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFFO1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16041-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This antibody works well in WB, IHC and IF applications.
This gene is a member of the intermediate filament family. Intermediate filaments are proteins which are primordial components of the cytoskeleton and nuclear envelope. The proteins encoded by the members of this gene family are evolutionarily and structurally related but have limited sequence homology, with the exception of the central rod domain. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.Spécification
IFFO1 | |
Polyclonal | |
Unconjugated | |
IFFO1 | |
HOM TES 103, IFFO, IFFO1, Tumor antigen HOM TES 103 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
25900, 320678, 362437 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.52 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q0D2I5, Q8BXL9 | |
Iffo1 | |
IFFO1 Fusion Protein Ag8915 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu