Learn More
HPCAL4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 12470-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
May be involved in the calcium-dependent regulation of rhodopsin phosphorylation.Spécification
HPCAL4 | |
Polyclonal | |
Unconjugated | |
HPCAL4 | |
hippocalcin like 4, Hippocalcin like protein 4, HLP4, HPCAL4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51440 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9UM19 | |
HPCAL4 | |
HPCAL4 Fusion Protein Ag3153 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |