Learn More
HBM Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca:  Proteintech 19996-1-AP-20UL
 vedi altre versioni di questo prodotto
Codice prodotto. 16819484
  • 136.48CHF / 20µL
Solo null disponibili
Spedizione stimata: 24-07-2025
Aggiungi al carrello
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Descrizione
Descrizione
The antibody is specific to HBM.
HBM, also named as HBAP2, belongs to the globin family.
Specifica
Specifica
HBM
Polyclonal
Unconjugated
HBM
HBAP2, HBAZ, HBM, HBZ, HBZ2, Hemoglobin mu chain, Hemoglobin subunit mu, Hemoglobin subunit zeta, Hemoglobin zeta chain, hemoglobin, mu, hemoglobin, zeta, Mu globin, Zeta globin
Rabbit
Antigen Affinity Chromatography
RUO
3042
-20°C
Liquid
Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6B0K9
HBM
Peptide