Learn More
GPR137B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16496-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
GPRs are characterized by the presence of 7 transmembrane domains. G Protein-Coupled Receptor (GPCR) TM7SF1 is a member of this family (subfamily: orphan U) and its expression has been reported in adult kidney and heart, but not in fetal kidney.Spécification
GPR137B | |
Polyclonal | |
Unconjugated | |
GPR137B | |
GPR137B, TM7SF1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7107 | |
-20°C | |
Liquid |
Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O60478 | |
GPR137B | |
GPR137B Fusion Protein Ag9633 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |