Learn More
GPR126 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 17774-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The GPR126 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human GPR126. This antibody recognizes human, mouse, rat antigen. The GPR126 antibody has been validated for the following applications: IHC, ELISA analysis.
Spécification
GPR126 | |
Polyclonal | |
Rabbit | |
Primary | |
Human, Rat, Mouse |
Immunohistochemistry, ELISA | |
Unconjugated | |
150 μL | |
57211 | |
IgG |