Learn More
FKBP7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 12092-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The protein encoded by this gene belongs to the FKBP-type peptidyl-prolyl cis/trans isomerase (PPIase) family. Members of this family exhibit PPIase activity and function as molecular chaperones. A similar protein in mouse is located in the endoplasmic reticulum and binds calcium.Spécification
FKBP7 | |
Polyclonal | |
Unconjugated | |
FKBP7 | |
23 kDa FK506 binding protein, 23 kDa FKBP, FK506 binding protein 7, FKBP 23, FKBP 7, FKBP23, FKBP7, PPIase, PPIase FKBP7, Rotamase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14231, 295672, 51661 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O54998, Q9Y680 | |
FKBP7 | |
FKBP7 Fusion Protein Ag2733 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |