Learn More
DBR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16019-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The RNA lariat debranching enzyme, or DBR1, specifically hydrolyzes 2-prime-to-5-prime branched phosphodiester bonds at the branch point of excised lariat intron RNA and converts them into linear molecules.Spécification
DBR1 | |
Polyclonal | |
Unconjugated | |
DBR1 | |
DBR1, Lariat debranching enzyme | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51163, 681234, 83703 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q923B1, Q9UK59 | |
DBR1 | |
DBR1 Fusion Protein Ag8830 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |