Learn More
CLTC Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 26523-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The CLTC antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human CLTC. This antibody recognizes human, pig, mouse, rat antigen. The CLTC antibody has been validated for the following applications: IF, IHC, IP, WB, ELISA analysis.
Spécification
CLTC | |
Polyclonal | |
Unconjugated | |
20 μL | |
1213 | |
IgG |
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence | |
300 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse, Rat, Pig |