Learn More
CD16 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16559-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The CD16 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CD16. This antibody recognizes human antigen. The CD16 antibody has been validated for the following applications: FC, IHC, WB, ELISA analysis.
Spécification
CD16 | |
Polyclonal | |
Unconjugated | |
150 μL | |
2214 | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry | |
200 μg/mL | |
Rabbit | |
Primary | |
Human |