Learn More
Agilent BioTek Cell Culture/Petri Dish Adapter for Imaging Multi-Mode Reader
For use with Imaging Multi-Mode Readers.
445.00CHF - 453.00CHF
Specifications
Product Type | Cell Culture/Petri Dish Adapter |
---|---|
For Use With (Equipment) | Imaging Multi-Mode Reader |
Product Code | Brand | Product Type | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Diameter (Metric) | Price | Quantity & Availability | ||||
15428722
|
Agilent BioTek
1222240 |
Cell Culture/Petri Dish Adapter | Dish diameter: 38.7 mm |
453.00CHF
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||
15458722
![]() |
Agilent BioTek
1222246 |
Cell Culture/Petri Dish Adapter | Dish diameter: 35.5 mm |
445.00CHF
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||
Description
For use with Imaging Multi-Mode Readers.Specifications
Cell Culture/Petri Dish Adapter | |
Imaging Multi-Mode Reader |