Learn More
Arfaptin-1 Mouse anti-Human, Unconjugated, Clone: 2A10B3, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 67461-1-IG-20UL
Description
The Arfaptin-1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Arfaptin-1. This antibody recognizes human, mouse, rat antigen. The Arfaptin-1 antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.
Spécification
Arfaptin-1 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry | |
2A10B3 | |
Unconjugated | |
20 μL | |
27236 | |
IgG2a |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.