Learn More
Ansell™ Crusader™ Flex Gloves
Non-woven felt insulation and a full-length heat barrier designed for intermittent handling of objects up to 400°F. Ansell™ Crusader™ Flex Gloves can be used in industries such as food service, and glass and metal fabrication.
22.65€
Product Code | Brand | dimSize | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | dimSize | Price | Quantity & Availability | |||||
11733994
|
Ansell™
42-445_9 |
N/A | |||||||
Description
- Front-and-back heat-insulated styles
- Provides protection from radiant heat and intermittent back-of-hand contact with hot objects
- Provides excellent grip when handling dry or oily surfaces
- Gauntlet slip-on cuff
- Contains no asbestos
- Length: 10 in. (25.4cm)
Not a replacement for silicate gloves. Designed for intermittent handling of objects up to 400°F (204°C).