Learn More
Sigma Laborzentrifugen™ Angle Rotors for Sigma™ Series 4 and 6 Centrifuges
Use these rotors with maximum load and at maximum speed for up to 50,000 cycles with low maintenance. Sigma™ Angle Rotors for Sigma™ Series 4 and 6 Centrifuges are ideal for use in research laboratories.
Brand:  Sigma Laborzentrifugen™ 12500
 View more versions of this product
Product Code. 11700679
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Fixed-angle rotors are used for sedimentation with high centrifugal forces of up to and more than 60,000 x g
  • Allow shorter sedimentation times in comparison to swing-bucket rotors 
  • Higher rotational speeds and higher centrifugal forces can be achieved
  • Sedimentation distance is also generally shorter than with swing-bucket rotors 
  • Capacity of the rotor chamber can be better utilized than with swing-bucket rotors
  • Rotors can be operated continuously at maximum speed and +4°C in nearly all Sigma centrifuges
Specifications
Specifications
Angle Rotor
8,000/9,600 rpm
30°
6 x 500 mL
Sigma 6-16, 6-H, 6-16K, 6-16KH, 6-16KR