Learn More
Sigma Laborzentrifugen™ Angle Rotors for Sigma™ Series 4 and 6 Centrifuges
Use these rotors with maximum load and at maximum speed for up to 50,000 cycles with low maintenance. Sigma™ Angle Rotors for Sigma™ Series 4 and 6 Centrifuges are ideal for use in research laboratories.
1725.00 € - 9520.00 €
Specifications
Type Angle Rotor
View More Specs
Products 10
Product Code Brand Capacity Max. Speed For Use With (Equipment) Price Quantity & Availability  
Product Code Brand Capacity Max. Speed For Use With (Equipment) Price Quantity & Availability  
10698792
View Documents
Sigma Laborzentrifugen™
12200
100 x 1.5/2.2 mL 9,000/9,600 rpm Sigma 4-16, 4-16K, 4-16KH, 4-16KR, 6-16, 16K, 16KH, 16KR This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10438621
View accessory products View Documents
Sigma Laborzentrifugen™
12172
12 x 30 mL 12,500/14,000 rpm Sigma 4-16, 4-16K, 4-16KH, 4-16KR, 4-16S, 4-16KS, 6-16, 6-16K
2955.00 €
Each
10739412
View accessory products View Documents
Sigma Laborzentrifugen™
12130
30 x 1.5/2.2 mL 13,500 rpm Sigma 4-16, 4-16K, 4-16KH, 4-16KR, 6-16, 6-16K
1725.00 €
Each
13465185
View Documents
Sigma Laborzentrifugen™
12449
48 x 1.5/2 mL 13,500/15,000 rpm Sigma 4-16S and 4-16KS
2200.00 €
Each
Only null left
Estimated Shipment: 22-07-2025
Add to basket
13455185
View Documents
Sigma Laborzentrifugen™
12356
6 x 250 mL 8,000/9,000 rpm Sigma 4-16S and 4-16KS
4605.00 €
Each
11700679
View accessory products View Documents
Sigma Laborzentrifugen™
12500
6 x 500 mL 8,000/9,600 rpm Sigma 6-16, 6-H, 6-16K, 6-16KH, 6-16KR This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10259771
View Documents
Sigma Laborzentrifugen™
12166
8 x 50 mL 12,500/14,000 rpm Sigma 4-16, 4-16K, 4-16KH, 4-16KR, 6-16, 6-16K
1970.00 €
Each
13475185
View Documents
Sigma Laborzentrifugen™
12600
6 x 500 mL 8,000/9,600 rpm 6-16S and 6-16KS
9520.00 €
Each
13435185
View Documents
Sigma Laborzentrifugen™
12266
8 x 50 mL 12,500/14,000 rpm Sigma 4-16S and 4-16KS
1965.00 €
Each
10535922
View accessory products View Documents
Sigma Laborzentrifugen™
12169
8 x 50 mL 12,500/13,500 rpm Sigma 4-16, 4-16K, 4-16KH, 4-16KR, 6-16, 6-16K
2685.00 €
Each
Description
Description
  • Fixed-angle rotors are used for sedimentation with high centrifugal forces of up to and more than 60,000 x g
  • Allow shorter sedimentation times in comparison to swing-bucket rotors 
  • Higher rotational speeds and higher centrifugal forces can be achieved
  • Sedimentation distance is also generally shorter than with swing-bucket rotors 
  • Capacity of the rotor chamber can be better utilized than with swing-bucket rotors
  • Rotors can be operated continuously at maximum speed and +4°C in nearly all Sigma centrifuges
Specifications
Specifications
Angle Rotor