Learn More
ABLIM3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27981-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The LIM domain is a double zinc finger structure that promotes protein-protein interactions. LIM domain proteins, such as ABLIM3, play roles in embryonic development, cell lineage determination, and cancer (Krupp et al., 2006 ).Spécification
ABLIM3 | |
Polyclonal | |
Unconjugated | |
ABLIM3 | |
abLIM 3, ABLIM3, Actin binding LIM protein 3, HMFN1661, KIAA0843 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22885, 319713 | |
-20°C | |
Liquid |
Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O94929, Q69ZX8 | |
Ablim3 | |
ABLIM3 Fusion Protein Ag27578 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |