Learn More
ENO1/2/3 Rabbit, anti-Human, CoraLite Plus 488, Clone: 6E+24, Proteintech
Rabbit Recombinant Monoclonal Antibody
Marque:  Proteintech CL488-81504-100UL
 Afficher plus de versions de ce produit
Code produit. 18057295
  • 450.00€ / 100µL
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
The ENO1/2/3 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human ENO1/2/3. This antibody recognizes human, mouse, rat, pig antigen. The ENO1/2/3 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Spécification
Spécification
ENO1/2/3
1000 μg/mL
Alpha enolase, C myc promoter binding protein, ENO1, ENO1L1, Enolase 1, enolase 1, (alpha), MBP 1, MBPB1, MPB 1, MPB1, NNE, Non neural enolase, Phosphopyruvate hydratase, Plasminogen binding protein, PPH
100 μL
2023
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence
CoraLite Plus 488
Rabbit
Primary
Human, Mouse, Rat, Pig
150 kDa