Learn More
ENO1/2/3 Rabbit, anti-Human, CoraLite Plus 488, Clone: 6E+24, Proteintech
Rabbit Recombinant Monoclonal Antibody
Marque: Proteintech CL488-81504-100UL
Description
The ENO1/2/3 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human ENO1/2/3. This antibody recognizes human, mouse, rat, pig antigen. The ENO1/2/3 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Spécification
ENO1/2/3 | |
1000 μg/mL | |
Alpha enolase, C myc promoter binding protein, ENO1, ENO1L1, Enolase 1, enolase 1, (alpha), MBP 1, MBPB1, MPB 1, MPB1, NNE, Non neural enolase, Phosphopyruvate hydratase, Plasminogen binding protein, PPH | |
100 μL | |
2023 | |
Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
Liquid |
Immunofluorescence | |
CoraLite Plus 488 | |
Rabbit | |
Primary | |
Human, Mouse, Rat, Pig | |
150 kDa | |
IgG |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.