Learn More
Saint Gobain Life Sciences Pure-Fit Tube Clamps PVDF
Pure-Fit TC's innovative design allows for complete flow stoppage, and the clamp can be installed over the tube in the normal fashion, or put together after the tube is already part of a completed assembly.
Marque:  Saint Gobain Life Sciences PFTC750PVDFUA
 Afficher plus de versions de ce produit
Code produit. 30040234
  • 4775.00€ / 250 pièces
Il en reste null
Expédition estimée: 22-09-2025
Ajouter au panier
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
Pure-Fit™ TC tube clamps represent a new generation of clamping technology for the pharmaceutical industry. They feature an outer body with no sharp edges, eliminating the potential for snags, scratches, or punctures; a press-down locking system that is fully encapsulated to guard against untimely or unwanted release; a side release mechanism that facilitates secure, single-handed installation and operation; and an integral bore hole for absolute lock-out capabilities.
  • Provides complete fluid stoppage
  • Smooth edges and corners help prevent punctures and ruptures
  • Can be installed on or over tubing or finished assemblies
  • High degree of leverage accommodates tubing with different durometers
  • Single-hand installation capability for ease of operation
  • Unique top locking mechanism and side release prevent unwanted openings
  • Available in PVDF or polypropylene
  • Fully autoclavable and sterilizable
  • Custom colors available
Applications
  • Pharmaceutical Processes
  • Peristaltic Pump Sets
  • Drug Delivery and Discovery
  • Medical Systems
  • Laboratory Functions
  • Assemblies and Tubing Sets
TRUSTED_SUSTAINABILITY
Spécification
Spécification
Autoclavables
Tubing Clamp
Clear
3/8 to 3/4 in.
250/Pk.
PVDF
Standard Taper