Learn More
Kova Plastics by Alltrista Lame de 10 cellules Glasstic™ avec grille
Use optically clear plastic for microscopic analysis with same refractive index as glass. Glasstic™ Slide 10 with Grids is a 10-chambered slide with a hemocytometer-type grid.
Marque: Kova Plastics by Alltrista 22270141
Description
Use optically clear plastic for microscopic analysis with same refractive index as glass. Glasstic™ Slide 10 with Grids is a 10-chambered slide with a hemocytometer-type grid.
Glasstic Slides sets the standard for accurate, reproducible, cost-effective and time-efficient standardized microscopic analysis. Glasstic Slides are extremely time-efficient and cost-effective.
- Manufactured from optically clear plastic with same refractive index as glass
- Each 10-chambered Glasstic Slide features 10 individual fully segregated specimen wells
- Capillary action reduces pipetting errors
- Suspends sediments homogeneously throughout the specimen
- Eliminates other variables, providing precise, reproducible fluid distribution throughout each chamber
- Chamber walls prevent specimen leakage and cross-contamination
- Glasstic slide grid is in the focal plane of the specimen, allowing for use over a range of magification
- Provides results that have high correlation with results obtained with precision hemocytometers: red blood cells: r: 0.99; white blood cells: r: 0.99; platelets: r: 0.93
Spécification
100 lames Glasstic 10 lames avec grilles | |
Oui | |
10 |
Examen microscopique de l’urine et d’autres liquides organiques | |
En plastique avec les qualités optiques du verre | |
L’unité |
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.