Learn More
Afora™ Vacuum Flask
Aspirate and collect fluids using the Afora™ Vacuum Flask as a reliable source.
124.00€ - 427.00€
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11911819
|
Afora™
45/E |
1000 mL |
124.00€
Each |
Estimated Shipment: 21-07-2025
Log in to see stock. |
|||||
11951779
|
Afora™
43/100 |
100 mL | N/A | ||||||
11921859
|
Afora™
46 |
2000 mL | N/A | ||||||
11951809
|
Afora™
44/E |
500 mL | N/A | ||||||
11981779
|
Afora™
43/E |
250 mL | N/A | ||||||
Description
- Glass
- Vacuum capability
Specifications
Vacuum |