Learn More
TTPA Mouse anti-Human, Unconjugated, Clone: 3E7D10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67431-1-IG-150UL
 View more versions of this product
Product Code. 16870056
  • 410.00€ / 150µL
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Description
Description
The TTPA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TTPA. This antibody recognizes human, mouse, rat, pig antigen. The TTPA antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
TTPA
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
3E7D10
Unconjugated
150 μL
7274
IgG2b