missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAPPC9 Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16014-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
NIBP is a 1246 amino acid intracellular regulatory protein. Reports suggest that NIBP physically interacts with NIK, IKK (beta), but not IKK (alpha) or IKK (gamma) and thus acts as a novel enhancer of cytokine-induced NF-(kappa) B signaling pathway. Due to its neuronal expression NIBP is considered as a potential target for modulating the NF-(kappa) B signaling cascade in neuronal pathologies dependent upon abnormal activation of this pathway. NIBP is mainly expressed in brain, muscle, heart, and kidney.Specifica
TRAPPC9 | |
Polyclonal | |
Unconjugated | |
TRAPPC9 | |
IBP, IKBKBBP, KIAA1882, NIBP, NIK and IKBKB binding protein, T1, TRAPPC9, TRS120, Tularik gene 1 protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100334971, 315059, 76510, 83696 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunoprecipitation | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3U0M1, Q96Q05 | |
Trappc9 | |
TRAPPC9 Fusion Protein Ag8791 | |
20 μL | |
Primary | |
Mouse, Human, Rat, Zebrafish | |
Antibody | |
IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto