Learn More
TIMM44 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13859-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
TIMM44 | |
Polyclonal | |
Unconjugated | |
TIMM44 | |
MIMT44, TIM44, TIMM44 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10469, 21856, 29635 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot, Immunoprecipitation | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O35094, O35857, O43615 | |
TIMM44 | |
TIMM44 Fusion Protein Ag4834 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |