Learn More
TBX22 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 11133-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This gene is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. Mutations in this gene have been associated with the inherited X-linked disorder, Cleft palate with ankyloglossia, and it is believed to play a major role in human palatogenesis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Spécification
TBX22 | |
Polyclonal | |
Unconjugated | |
TBX22 | |
CLPA, T box 22, T box protein 22, TBOX22, TBX22, TBXX | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
245572, 302369, 50945 | |
-20°C | |
Liquid |
Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8K402, Q9Y458 | |
TBX22 | |
TBX22 Fusion Protein Ag1591 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |