Learn More
Fisherbrand™ Stirring Spatulas
Fisherbrand™ Stirring Spatulas can be used for a variety of applications such as mixing samples and solutions in test tubes.
Brand:  Fisherbrand™ 9409501
Encompass_Preferred
Product Code. 10292561
  • 9.00 € / Pack of 500
Only null left
In Stock
Description
  • Polystyrene construction
  • Transparent
  • Applications: mixing samples and solutions in test tubes, adding small reagent volumes and releasing blood clots from the side of test tube walls
Specifications
Specifications
500 Pk.
3 mm
Polystyrene
120 mm