Learn More
SRF Mouse anti-Human, CoraLite 594, Clone: 1G8F4, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech CL594-66742-100UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The SRF antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SRF. This antibody recognizes human antigen. The SRF antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Spécification
SRF | |
Monoclonal | |
CoraLite 594 | |
100 μL | |
6722 | |
IgG2a |
Immunofluorescence | |
1G8F4 | |
Mouse | |
Primary | |
Human |