Learn More
Bel Art™ Poxygrid™ Rack and a Half
Extra-long wire test tube racks for high volume test tube work
Brand: Bel Art™ F18786-0780
60.82 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
This item is not returnable.
View return policy
Description
- High quality epoxy-coated wire racks
- Epoxy coated steel wire is chemical and corrosion resistant
- Steam autoclavable at 121°C (250°F)
Specifications
Test Tube Rack | |
Autoclavable | |
Tubes (13 to 16 mm) | |
100 | |
3 Tiers | |
Non-sterile | |
-223°C to +121°C | |
16 mm |
5 x 20 | |
Blue | |
Epoxy-coated Steel | |
Rectangular | |
Yes, When Empty | |
13 to 16mm Tubes | |
38.6 x 10.8 x 6.4 cm |