missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloride, Free-flowing, Reagent Grade, anhydrous, ≥99%, Honeywell Fluka™

Code produit. 15647120 Tous les produits Honeywell Fluka Produits
5 kg, Flacon poly, 5kg
missing translation for 'orderingAttributeHoverText'
Quantité:
500 g
1 kg
2.5 kg
5 kg
12 kg
25 kg
Conditionnement:
Boîte en carton
Flacon poly
missing translation for 'unitSize'
12kg
1kg
2.50kg
25kg
500g
5kg
This item is not returnable. View return policy
This item is not returnable. View return policy

À écoulement libre, qualité réactif, anhydre, ≥ 99%

  • ≥95 % pour usage général en laboratoire.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-14-5
Formule moléculaire ClNa
Poids moléculaire (g/mol) 58.44
Numéro MDL MFCD00003477
Clé InChI FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonyme sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
CID PubChem 5234
ChEBI CHEBI:26710
SMILES [Na+].[Cl-]

Specifications

Point d’ébullition 1,465°C
Nom chimique ou matériau Sodium chloride
Point de fusion 801°C (literature)
Note relative au nom Free-flowing, Reagent Grade, anhydrous, ≥99%
% max. CAS ≤100.0000%
Quantité 5 kg
Formule linéaire NaCl
Beilstein 3534976
Nom IUPAC sodium;chloride
Poids de la formule 58.44
Pourcentage de pureté ≥99%
Qualité Réactif
Conditionnement Flacon poly
Densité 2.160 g/cm3
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium chloride, Free-flowing, Reagent Grade, anhydrous, ≥99%, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.