Learn More
BRAND™ Robotic Tips, BIO-CERT™ Liquid Handling, Sterile
Pipette tips offer reliable quality for precise and demanding analyses with the pipetting robot Liquid Handling Station.
Brand: BRAND™ 734168
83.30 EUR valid until 2025-12-31
Use promo code "24288" to get your promotional price.
This item is not returnable.
View return policy
Description
- The raw materials are free from DiHEMDA und Oleamid. Tips are manufactured at BRAND under monitored cleanroom conditions and are automatically packaged. No parting agents or demolding aids are used. All racked tips and filter tips up to 1000 μL are free of DNA (< 40 fg), RNase (< 8.6 fg), endotoxins (< 1 pg) and ATP (< 1 fg).
- For use with pipetting robots, tips are required that are matched to the robot's mode of operation in terms of geometry and surface properties. BRAND robotic pipette tips and robotic filter tips meet all requirements for use with the Liquid Handling Station pipetting robot.
Specifications
10 to 200 μL | |
Sterile | |
Robotic Tips |
Filtered | |
TipRack (960/Pk.) | |
Polypropylene |