Learn More
Labconco™ Replacement Freeze Dry Tubing
Neoprene construction. Labconco™ Replacement Freeze Dry Tubing allows moisture from condensation to be emptied from the freeze dryer.
Marque: Labconco™ 7645900
Description
- For Dry Ice Benchtop Freeze Dry System connections or as drain hose on FreeZone 6, 12, and 18 Liter Freeze Dryers manufactured before August 2017
- Replacement drain hose allows moisture from condensation to be emptied from the freeze dryer
- 8mm (5/16 in.) ID
- Length: 0.9m (3 ft.)
- Verify that the part matches your FreeZone Freeze Dryer's volume before purchasing
Spécification
Rubber | |
3 ft. | |
Semirigid |