Learn More
Helios Italquartz Rectangular Basins/Dishes, Transparent
Rectangular basins/dishes constructed of transparent, durable, resistant, and chemically inert quartz glass.
Brand:  Helios Italquartz 62P00205
127.31 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15808044
  • 179.00€ / Each
Only null left
Estimated Shipment: 18-09-2025
Description
  • Transparent
  • Rectangular form
  • Chemically inert
  • Excellent electrical insulation
  • Excellent strength and durability
  • Exceptional thermal stability compared to glass
Specifications
Specifications
Transparent
Rectangular
68 x 12 mm
12 mm
Quartz
Dish
68/39 mm
12 mm