Learn More
Macherey-Nagel™ Potassium Iodide Starch Paper
Impregnated with potassium iodide and starch for the detection of nitrite and free chlorine. Macherey Nagel™ Potassium Iodide Starch Paper may be used by dipping it into the solution to be tested.
Brand: Macherey-Nagel™ 90756
10.00 EUR valable jusqu'au 2025-12-31
Utilisez le code promo "25339" pour bénéficier de cette offre.
Description
- Grade MN 816 N
- Normal sensitivity
- Spot-test by applying drops
- Also used to control diazotization reactions
Specifications
Bande | |
100/paquet | |
Papier amidonné d’iodure de potassium | |
75 x 10 mm |