Learn More
mb2 Swivel Antistatic Work Chair in Polyurethane
Click to view available options
Holds:
Patins
Unit Size:
Each
Description
- Antistatic work chair
- Seat and backrest in multilayers of beech and poplar wood, entirely foamed with black antistatic polyurethane
- Backrest support in chromed steel rod, in height and depth adjustable
- Gas height adjustment
- Antistatic glides
- 5-spoke base in die-cast polished aluminium
Specifications
Specifications
Product Type | ESD Chair |
Color | Black |
Adjustable (Height) | Yes |
Adjustable | Height |
Autoclavable | Not Autoclavable |
Back Style | Standard |
Height (Metric) | 870 mm |
Holds | Patins |
Quantity | 1 Ea. |
Seating Height | 540 to 480 mm |