Learn More
PLCG2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27173-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The PLCG2 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human PLCG2. This antibody recognizes human antigen. The PLCG2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spécification
PLCG2 | |
Polyclonal | |
Unconjugated | |
20 μL | |
5336 | |
IgG |
Western Blot, Immunohistochemistry, ELISA | |
400 μg/mL | |
Rabbit | |
Primary | |
Human |