Learn More
Poudre de noir de platine, HiSPEC 1000, Thermo Scientific Chemicals
Code produit. 11377537
Click to view available options
Quantité:
0.25 g
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
CAS: 7440-06-4 | Pt | 195.08 g/mol
Platinum black is most commonly used for electrolyte fuel cell, microbial fuel cell, polymer electrolyte fuel cell and phosphoric acid fuel cells. It is used as a catalyst in hydrogenation reactions and a gas adsorbent. It can be coated on electrode surface by electroplating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 7440-06-4
Formule moléculaire Pt
Poids moléculaire (g/mol) 195.08
Numéro MDL MFCD00011179
Clé InChI BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonyme black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CID PubChem 23939
ChEBI CHEBI:33400
Nom IUPAC platine
SMILES [Pt]
Spécification
Nom chimique ou matériau Platinum black
Aspect Dark gray powder
Densité 21.45g/cm3 at 20°C
Quantité 5 g
Informations sur la solubilité Platinum black is insoluble in water and soluble in aqua regia.
Odeur Odorless
Forme Poudre