Learn More
PHF14 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24787-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
PHF14 gene ontology annotations related to this gene include metal ion binding.Spécification
PHF14 | |
Polyclonal | |
Unconjugated | |
PHF14 | |
KIAA0783, PHD finger protein 14, PHF14 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9678 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O94880 | |
PHF14 | |
PHF14 Fusion Protein Ag20542 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |