Learn More
Thermo Scientific™ Oxoid™ Malt Extract
Oxoid Malt Extract is recommended for use in media for yeast and mold growth.
595.00 NOK
Specifications
Form Powder
Product Type Malt Extract
View More Specs
Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10188572
View Documents Promotion Details
Thermo Scientific™
LP0039B
500 g
595.00 NOK
500g
Description
Description
Oxoid Malt Extract is recommended for use in media for yeast and mold growth.
An extract of sprouted grain with conserved nitrogen and carbohydrate constituents
Specifications
Specifications
Powder
Malt Extract
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Malt Extract
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304-IF INHALED:.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : Barley, malt