missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ omniPAGE WAVE Maxi Vertical Electrophoresis Modular System
Shop All Fisherbrand Products
Click to view available options
No. of Gels:
2 to 4
Unit Size:
Each
Description
- Includes all modules and accessories required for Slab Gel Electrophoresis, 2-D Electrophoresis and Electroblotting
- Rapid and intuitive casting system
- Enhanced set up cooling system
- Increased capacity can run up to four gels per run
Specifications
Specifications
Cooling Type | Detachable cooling coils |
Type | Vertical Electrophoresis System |
Gel Dimensions | W x H: 18.8 x 17.5 cm (7.4 x 6.89 in.) |
Includes | 1x WAVE Maxi Vertical Unit which includes: PAGE insert, 2x4mm thick notched glass plates, Plain glass plates (2 x 4mm thick), Bonded spacers (1mm thick), 1 x dummy plate, 2 x combs (1mm thick 24 samples), 1 x casting base, silicone mat, cooling coil, 1 x WAVE Standard Electroblotting Module which includes: WAVE Maxi Platinum Wire Blotting insert, 4 x compression cassettes and 18 x fibre pads |
No. of Gels | 2 to 4 |
Volume (English) Buffer | 0.32 to 1.48 gal. |
Volume (Metric) Buffer | 1200 to 5600 mL |
Width (English) | 10.23 in. |
Width (Metric) | 26 cm |
Height (English) | 11.02 in. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction