Learn More
OFD1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 22851-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This antibody recognize all the isoforms of OFD1. The 43 kDa is isoform 2. The ∽80 kDa band is unknown.
This gene is located on the X chromosome and encodes a centrosomal protein. A knockout mouse model has been used to study the effect of mutations in this gene. The mouse gene is also located on the X chromosome, however, unlike the human gene it is not subject to X inactivation. Mutations in this gene are associated with oral-facial-digital syndrome type I and Simpson-Golabi-Behmel syndrome type 2. Many pseudogenes have been identified; a single pseudogene is found on chromosome 5 while as many as fifteen have been found on the Y chromosome.Spécification
OFD1 | |
Polyclonal | |
Unconjugated | |
OFD1 | |
71 7A, CXorf5, OFD1, oral facial digital syndrome 1, Protein 71 7A, SGBS2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8481 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75665 | |
OFD1 | |
OFD1 Fusion Protein Ag18898 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |